siRNA Details
| Gene Symbol: RASGEF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300735.1 | NP_001287664.1 | 7 | 915 | ||
| NM_001300736.1 | NP_001287665.1 | 6 | 792 | |||
| NM_152545.2 | NP_689758.1 | 7 | 918 | |||
| XM_017007812.1 | XP_016863301.1 | 5 | 636 | |||
| XM_017007813.1 | XP_016863302.1 | 7 | 1006 | |||
| GenBank mRNA | AK056257.1 | - | 7 | 897 | ||
| AK291459.1 | - | 7 | 863 | |||
| AK303625.1 | - | 6 | 767 | |||
| BC121003.1 | - | 7 | 830 | 40116919 | ||
| BC121004.1 | - | 6 | 707 | 40116920 | ||

RASGEF1B
Genomic Map