siRNA Details
| Gene Symbol: TMEM18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152834.2 | NP_690047.2 | 5 | 1730 | ||
| GenBank mRNA | AL137269.1 | - | 5 | 1730 | ||
| BC032379.1 | - | 5 | 1990 | 5109444 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM18| Gene Symbol: TMEM18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152834.2 | NP_690047.2 | 5 | 1730 | ||
| GenBank mRNA | AL137269.1 | - | 5 | 1730 | ||
| BC032379.1 | - | 5 | 1990 | 5109444 | ||