siRNA Details
| Gene Symbol: ZNF548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172773.1 | NP_001166244.1 | 4 | 2076 | ||
| NM_152909.3 | NP_690873.2 | 3 | 2040 | |||
| XM_017026316.1 | XP_016881805.1 | 6 | 2346 | |||
| GenBank mRNA | AK299069.1 | - | 4 | 1923 | ||
| BC030788.2 | - | 3 | 2040 | 5266647 | ||

ZNF548
Genomic Map