siRNA Details
| Gene Symbol: WDFY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020945.1 | NP_065996.1 | 14, 15 | 2840 | ||
| XM_005270004.3 | XP_005270061.1 | 14, 15 | 2955 | |||
| XM_011539986.2 | XP_011538288.2 | 14, 15 | 2958 | |||
| XM_011539987.2 | XP_011538289.2 | 14, 15 | 2961 | |||
| XM_011539988.2 | XP_011538290.1 | 15, 16 | 3039 | |||
| XM_011539990.2 | XP_011538292.1 | 14, 15 | 2986 | |||
| XM_011539991.2 | XP_011538293.1 | 14, 15 | 2990 | |||
| XM_011539992.2 | XP_011538294.1 | 14, 15 | 2990 | |||
| XM_017016463.1 | XP_016871952.1 | 15, 16 | 2882 | |||
| XM_017016465.1 | XP_016871954.1 | 14, 15 | 2967 | |||
| GenBank mRNA | AK127650.1 | - | 14, 15 | 2849 | ||

WDFY4
Genomic Map