siRNA Details
| Gene Symbol: ZNF547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173631.3 | NP_775902.2 | 4 | 1135 | ||
| GenBank mRNA | AK055662.1 | - | 4 | 1047 | ||
| AK291464.1 | - | 4 | 982 | |||
| AL832545.1 | - | 4 | 1003 | |||
| BC042681.1 | - | 4 | 1004 | 4823926 | ||

ZNF547
Genomic Map