siRNA Details
| Gene Symbol: RABL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173825.3 | NP_776186.2 | 8 | 982 | ||
| XM_006713595.2 | XP_006713658.1 | 7 | 894 | |||
| XM_017006227.1 | XP_016861716.1 | 9 | 1008 | |||
| XM_017006228.1 | XP_016861717.1 | 8 | 936 | |||
| GenBank mRNA | AK025772.1 | - | 5 | 678 | ||
| AK293085.1 | - | 8 | 966 | |||
| BC020832.1 | - | 8 | 961 | 4771781 | ||
| BX647559.1 | - | 8 | 979 | |||
| BX647593.1 | - | 8 | 982 | |||
| BX647664.1 | - | 8 | 966 | |||

RABL3
Genomic Map