siRNA Details
| Gene Symbol: KRTCAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321325.1 | NP_001308254.1 | 7 | 843 | ||
| NM_173853.3 | NP_776252.2 | 7 | 846 | |||
| GenBank mRNA | AY157576.1 | - | 7 | 816 | ||
| AY358993.1 | - | 6 | 901 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTCAP3| Gene Symbol: KRTCAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321325.1 | NP_001308254.1 | 7 | 843 | ||
| NM_173853.3 | NP_776252.2 | 7 | 846 | |||
| GenBank mRNA | AY157576.1 | - | 7 | 816 | ||
| AY358993.1 | - | 6 | 901 | |||