siRNA Details
| Gene Symbol: ACSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004457.3 | NP_004448.2 | 5 | 990 | ||
| NM_203372.1 | NP_976251.1 | 4 | 883 | |||
| XM_017003584.1 | XP_016859073.1 | 3 | 802 | |||
| GenBank mRNA | AB061712.1 | - | 5 | 958 | ||
| AK023191.1 | - | 2 | 535 | |||
| AK314621.1 | - | 5 | 968 | |||
| BC041692.1 | - | 5 | 982 | 5226905 | ||
| D89053.1 | - | 3 | 681 | |||

ACSL3
Genomic Map