siRNA Details
| Gene Symbol: SLC16A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323977.1 | NP_001310906.1 | 5 | 698 | ||
| NM_001323978.1 | NP_001310907.1 | 6 | 1126 | |||
| NM_001323979.1 | NP_001310908.1 | 5 | 813 | |||
| NM_001323980.1 | NP_001310909.1 | 5 | 1043 | |||
| NM_001323981.1 | NP_001310910.1 | 6 | 1256 | |||
| NM_194298.2 | NP_919274.1 | 5 | 1173 | |||
| XM_017015883.1 | XP_016871372.1 | 5 | 1173 | |||
| XM_017015884.1 | XP_016871373.1 | 6 | 1506 | |||
| GenBank mRNA | AK125791.1 | - | 5 | 833 | ||
| BC126238.1 | - | 4 | 762 | 8991954 | ||
| BC130567.1 | - | 4 | 762 | 40146598 | ||

SLC16A9
Genomic Map