siRNA Details
| Gene Symbol: ZNF542P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003127.2 | 7 | 1511 | |||
| NR_024055.2 | 6 | 1414 | ||||
| NR_024056.2 | 6 | 1667 | ||||
| NR_024057.2 | 5 | 1595 | ||||
| NR_033418.1 | 6 | 1735 | ||||
| GenBank mRNA | AK097128.1 | - | 5 | 1399 | ||
| AL833621.1 | - | 5 | 1364 | |||
| BC089419.1 | - | 5 | 1339 | |||
| BX640680.1 | - | 7 | 1471 | |||
| BX647919.1 | - | 6 | 1470 | |||

ZNF542P
Genomic Map