siRNA Details
| Gene Symbol: ZNF568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204835.1 | NP_001191764.1 | 7 | 2468 | ||
| NM_001204836.1 | NP_001191765.1 | 6 | 2210 | |||
| NM_001204837.1 | NP_001191766.1 | 5 | 2151 | |||
| NM_198539.3 | NP_940941.2 | 7 | 2471 | |||
| XM_017026773.1 | XP_016882262.1 | 8 | 2555 | |||
| XM_017026774.1 | XP_016882263.1 | 7 | 2160 | |||
| XM_017026779.1 | XP_016882268.1 | 7 | 3099 | |||
| XM_017026780.1 | XP_016882269.1 | 6 | 3023 | |||
| GenBank mRNA | BX640681.1 | - | 5 | 2135 | ||

ZNF568
Genomic Map