siRNA Details
Gene Symbol: BLOC1S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199322.1 | NP_001186251.1 | 6 | 1405 | ||
NM_001199323.1 | NP_001186252.1 | 4 | 1172 | |||
NM_201280.2 | NP_958437.1 | 5 | 1302 | |||
GenBank mRNA | AF426434.1 | - | 5 | 1302 | ||
AK025442.1 | - | 5 | 1256 | |||
BC032438.1 | - | 5 | 1266 | |||
BX647336.1 | - | 6 | 1475 | |||
CR749569.1 | - | 5 | 1277 |
Gene Symbol: EEF1E1-BLOC1S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037618.1 | 7 | 1610 |
