siRNA Details
| Gene Symbol: LRRC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201550.3 | NP_963844.2 | 1 | 1295 | ||
| GenBank mRNA | AK095935.1 | - | 1 | 318 | ||
| AK123908.1 | - | 1 | 891 | |||
| BC090881.1 | - | 1 | 1295 | 30530394 | ||
| BC106892.1 | - | 1 | 888 | |||
| BC106893.1 | - | 1 | 891 | 40030470 | ||

LRRC10
Genomic Map