siRNA Details
| Gene Symbol: LRRC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201550.3 | NP_963844.2 | 1 | 2317 | ||
| GenBank mRNA | AK095935.1 | - | 1 | 1340 | ||
| AK123908.1 | - | 1 | 1913 | |||
| BC090881.1 | - | 1 | 2317 | 30530394 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC10| Gene Symbol: LRRC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201550.3 | NP_963844.2 | 1 | 2317 | ||
| GenBank mRNA | AK095935.1 | - | 1 | 1340 | ||
| AK123908.1 | - | 1 | 1913 | |||
| BC090881.1 | - | 1 | 2317 | 30530394 | ||