siRNA Details
| Gene Symbol: FBXO33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203301.3 | NP_976046.1 | 4 | 3227 | ||
| GenBank mRNA | BC042535.1 | - | 2 | 1706 | ||
| BC053537.1 | - | 4 | 3144 | 5181903 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXO33| Gene Symbol: FBXO33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203301.3 | NP_976046.1 | 4 | 3227 | ||
| GenBank mRNA | BC042535.1 | - | 2 | 1706 | ||
| BC053537.1 | - | 4 | 3144 | 5181903 | ||