siRNA Details
| Gene Symbol: FHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243878.1 | NP_001230807.1 | 5 | 711 | ||
| NM_004468.4 | NP_004459.2 | 6 | 887 | |||
| XM_017000675.1 | XP_016856164.1 | 3 | 311 | |||
| GenBank mRNA | AF133732.1 | - | 5 | 708 | ||
| AK290641.1 | - | 6 | 887 | |||
| BC001351.2 | - | 6 | 877 | 2964682 | ||
| BC011697.2 | - | 6 | 877 | 3532981 | ||
| BC014501.1 | - | 5 | 698 | 4860329 | ||
| BT007052.1 | - | 5 | 708 | |||
| CR457425.1 | - | 5 | 708 | |||
| U60116.2 | - | 5 | 747 | |||

FHL3
Genomic Map