siRNA Details
Gene Symbol: ZNF321P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037805.1 | 2 | 830 | |||
GenBank mRNA | AK096828.1 | - | 2 | 830 | ||
BC046449.1 | - | 2 | 821 | 5186884 | ||
BC119627.1 | - | 2 | 829 | 40115508 | ||
BC119628.1 | - | 2 | 829 | 40115510 |
Gene Symbol: ZNF816-ZNF321P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202473.1 | NP_001189402.1 | 4 | 1176 | ||
GenBank mRNA | AK316503.1 | - | 4 | 1088 | ||
CR749224.1 | - | 4 | 1126 |
