siRNA Details
| Gene Symbol: ZNF321P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037805.1 | 2 | 1644 | |||
| GenBank mRNA | AK096828.1 | - | 2 | 1643 | ||
| BC046449.1 | - | 2 | 1636 | 5186884 | ||
| Gene Symbol: ZNF816-ZNF321P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202473.1 | NP_001189402.1 | 4 | 1990 | ||

ZNF321P
Genomic Map