siRNA Details
| Gene Symbol: CCDC187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291516.1 | NP_001278445.1 | 10 | 3127 | ||
| NR_111982.1 | 10 | 3541 | ||||
| XM_011518683.2 | XP_011516985.1 | 10 | 3449 | |||
| XM_011518684.2 | XP_011516986.1 | 9 | 3355 | |||
| GenBank mRNA | BC031926.1 | - | 10 | 3127 | ||
| BC042107.1 | - | 10 | 3517 | 5170879 | ||

CCDC187
Genomic Map