siRNA Details
| Gene Symbol: TBRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261834.1 | NP_001248763.1 | 2226 | |||
| NM_004749.3 | NP_004740.2 | 2270 | ||||
| NM_030900.3 | NP_112162.1 | 1940 | ||||
| NM_199122.2 | NP_954573.1 | 1919 | ||||
| GenBank mRNA | AF011792.1 | - | 1582 | |||
| AK222943.1 | - | 1876 | ||||
| AL833840.1 | - | 2239 | ||||
| BC002732.2 | - | 1709 | ||||
| BC014918.1 | - | 2200 | 3906670 | |||
| BC017235.2 | - | 2197 | 3538504 | |||

TBRG4
Genomic Map