siRNA Details
Gene Symbol: CNGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005140.1 | NP_005131.1 | 7 | 2614 | ||
GenBank mRNA | AK128186.1 | - | 7 | 2608 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CNGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005140.1 | NP_005131.1 | 7 | 2614 | ||
GenBank mRNA | AK128186.1 | - | 7 | 2608 |