siRNA Details
| Gene Symbol: CCDC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005436.4 | NP_005427.2 | 4, 5 | 916 | ||
| GenBank mRNA | AK292593.1 | - | 4, 5 | 916 | ||
| BC022284.1 | - | 4, 5 | 810 | |||
| BC036757.2 | - | 4, 5 | 755 | 5549623 | ||
| BC064391.1 | - | 4, 5 | 742 | 5745698 | ||

CCDC6
Genomic Map