siRNA Details
| Gene Symbol: PIBF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006346.2 | NP_006337.2 | 2 | 453 | ||
| XM_011534881.2 | XP_011533183.1 | 2 | 481 | |||
| XM_011534882.2 | XP_011533184.1 | 2 | 481 | |||
| XM_011534884.2 | XP_011533186.1 | 2 | 481 | |||
| XM_011534886.2 | XP_011533188.1 | 2 | 481 | |||
| XM_017020351.1 | XP_016875840.1 | 2 | 481 | |||
| XM_017020352.1 | XP_016875841.1 | 2 | 407 | |||
| XM_017020353.1 | XP_016875842.1 | 2 | 404 | |||
| XM_017020354.1 | XP_016875843.1 | 2 | 404 | |||
| GenBank mRNA | AF330046.1 | - | 2 | 495 | ||
| AK309828.1 | - | 2 | 410 | |||
| AY370776.1 | - | 2 | 352 | |||
| BC006009.1 | - | 2 | 406 | |||
| BC008432.1 | - | 2 | 406 | |||
| BC041806.1 | - | 2 | 405 | 5269898 | ||
| BC051911.1 | - | 2 | 383 | 6140514 | ||
| BX341357.2 | - | 2 | 453 | |||

PIBF1
Genomic Map