siRNA Details
| Gene Symbol: ZNF81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007137.3 | NP_009068.2 | 5 | 5119 | ||
| XM_005272600.3 | XP_005272657.1 | 5 | 5177 | |||
| XM_011543899.2 | XP_011542201.1 | 5 | 6783 | |||
| XM_011543900.2 | XP_011542202.1 | 6 | 5238 | |||
| XM_017029486.1 | XP_016884975.1 | 6 | 5296 | |||
| XM_017029487.1 | XP_016884976.1 | 5 | 5529 | |||
| GenBank mRNA | AK126339.1 | - | 1 | 2370 | ||
| AK126949.1 | - | 7 | 1054 | |||

ZNF81
Genomic Map