siRNA Details
| Gene Symbol: ZNF81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007137.3 | NP_009068.2 | 5 | 7432 | ||
| XM_005272600.3 | XP_005272657.1 | 5 | 7490 | |||
| XM_011543899.2 | XP_011542201.1 | 5 | 9096 | |||
| XM_011543900.2 | XP_011542202.1 | 6 | 7551 | |||
| XM_017029486.1 | XP_016884975.1 | 6 | 7609 | |||
| XM_017029487.1 | XP_016884976.1 | 5 | 7842 | |||
| GenBank mRNA | AK126949.1 | - | 7 | 3367 | ||
| BC111389.1 | - | 1 | 493 | |||
| DA316311.1 | - | 1 | 119 | |||

ZNF81
Genomic Map