siRNA Details
| Gene Symbol: ZNF770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014106.3 | NP_054825.2 | 3 | 770 | ||
| XM_011521744.2 | XP_011520046.1 | 2 | 614 | |||
| GenBank mRNA | AK000589.1 | - | 1 | 478 | ||
| AK291438.1 | - | 3 | 761 | |||
| BC071603.1 | - | 3 | 742 | 30340520 | ||
| BQ921117.1 | - | 1 | 196 | |||

ZNF770
Genomic Map