siRNA Details
| Gene Symbol: SECISBP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193489.1 | NP_001180418.1 | 4 | 829 | ||
| NM_014701.3 | NP_055516.2 | 4 | 829 | |||
| GenBank mRNA | AK307480.1 | - | 4 | 512 | ||
| AK308183.1 | - | 4 | 713 | |||
| BC033001.2 | - | 4 | 764 | 4825784 | ||
| D87445.3 | - | 4 | 699 | |||

SECISBP2L
Genomic Map