siRNA Details
| Gene Symbol: SECISBP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193489.1 | NP_001180418.1 | 5 | 1068 | ||
| NM_014701.3 | NP_055516.2 | 5 | 1068 | |||
| GenBank mRNA | AK307480.1 | - | 5 | 751 | ||
| BC033001.2 | - | 5 | 1003 | 4825784 | ||
| D87445.3 | - | 5 | 938 | |||

SECISBP2L
Genomic Map