siRNA Details
| Gene Symbol: DIP2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014974.2 | NP_055789.1 | 9 | 1181 | ||
| XM_005252426.3 | XP_005252483.1 | 10 | 1340 | |||
| XM_005252427.3 | XP_005252484.1 | 9 | 1221 | |||
| XM_005252428.4 | XP_005252485.1 | 10 | 1391 | |||
| XM_011519428.2 | XP_011517730.1 | 10 | 1322 | |||
| XM_011519429.2 | XP_011517731.1 | 10 | 1320 | |||
| XM_011519430.2 | XP_011517732.1 | 9 | 1197 | |||
| XM_011519431.2 | XP_011517733.1 | 9 | 1197 | |||
| XM_011519432.1 | XP_011517734.1 | 9 | 1622 | |||
| GenBank mRNA | AB023151.2 | - | 9 | 1179 | ||
| AK126063.1 | - | 10 | 1227 | |||
| AK310613.1 | - | 6 | 816 | |||
| BC063313.1 | - | 9 | 1181 | 30349267 | ||

DIP2C
Genomic Map