siRNA Details
| Gene Symbol: KLHDC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014997.3 | NP_055812.1 | 3 | 414 | ||
| XM_005250233.4 | XP_005250290.1 | 2 | 303 | |||
| GenBank mRNA | BC036464.2 | - | 3 | 396 | 4828273 | |
| BC044884.1 | - | 3 | 416 | 5273087 | ||
| D87454.1 | - | 3 | 156 | |||

KLHDC10
Genomic Map