siRNA Details
| Gene Symbol: FAM179B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015091.3 | NP_055906.2 | 13, 14 | 4394 | ||
| GenBank mRNA | AB007883.2 | - | 13, 14 | 4376 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM179B| Gene Symbol: FAM179B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015091.3 | NP_055906.2 | 13, 14 | 4394 | ||
| GenBank mRNA | AB007883.2 | - | 13, 14 | 4376 | ||