siRNA Details
| Gene Symbol: RPRD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297673.1 | NP_001284602.1 | 5 | 1170 | ||
| NM_001297674.1 | NP_001284603.1 | 5 | 1170 | |||
| NM_015203.4 | NP_056018.2 | 6 | 1248 | |||
| XM_005245033.2 | XP_005245090.1 | 6 | 1211 | |||
| XM_006711240.2 | XP_006711303.1 | 6 | 1211 | |||
| XM_011509344.2 | XP_011507646.1 | 6 | 1211 | |||
| XM_017000763.1 | XP_016856252.1 | 5 | 1133 | |||
| XM_017000764.1 | XP_016856253.1 | 6 | 586 | |||
| XM_017000765.1 | XP_016856254.1 | 6 | 1211 | |||
| XM_017000766.1 | XP_016856255.1 | 5 | 845 | |||
| XM_017000767.1 | XP_016856256.1 | 5 | 1133 | |||
| GenBank mRNA | AB007929.2 | - | 6 | 617 | ||
| AK056707.1 | - | 6 | 655 | |||
| AK125864.1 | - | 6 | 818 | |||
| AK291703.1 | - | 6 | 666 | |||
| AK304380.1 | - | 5 | 588 | |||
| AK308148.1 | - | 5 | 579 | |||
| BC045623.1 | - | 5 | 571 | 5269891 | ||
| BX641025.1 | - | 7 | 1143 | |||

RPRD2
Genomic Map