siRNA Details
| Gene Symbol: MED13L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015335.4 | NP_056150.1 | 2 | 146 | ||
| XM_011538080.2 | XP_011536382.1 | 2 | 179 | |||
| XM_011538081.2 | XP_011536383.1 | 2 | 179 | |||
| XM_011538082.2 | XP_011536384.1 | 2 | 349 | |||
| XM_017019090.1 | XP_016874579.1 | 2 | 179 | |||
| GenBank mRNA | AF515599.1 | - | 2 | 146 | ||
| AY338463.1 | - | 2 | 298 | |||
| BC130422.1 | - | 2 | 142 | 40146453 | ||

MED13L
Genomic Map