siRNA Details
| Gene Symbol: MED13L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015335.4 | NP_056150.1 | 6 | 828 | ||
| XM_011538080.2 | XP_011536382.1 | 6 | 861 | |||
| XM_011538081.2 | XP_011536383.1 | 6 | 861 | |||
| XM_011538082.2 | XP_011536384.1 | 6 | 1031 | |||
| XM_017019090.1 | XP_016874579.1 | 6 | 861 | |||
| GenBank mRNA | AF515599.1 | - | 6 | 828 | ||
| AK127706.1 | - | 3 | 362 | |||
| AY338463.1 | - | 6 | 980 | |||
| BC130422.1 | - | 6 | 824 | 40146453 | ||
| CR749332.1 | - | 3 | 352 | |||

MED13L
Genomic Map