siRNA Details
| Gene Symbol: KBTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015483.2 | NP_056298.2 | 4 | 3398 | ||
| XM_005249696.2 | XP_005249753.1 | 4 | 3254 | |||
| XM_011515256.2 | XP_011513558.1 | 4 | 3133 | |||
| GenBank mRNA | AB040922.1 | - | 1 | 4022 | ||
| AF004292.1 | - | 1 | 757 | |||
| AF161402.1 | - | 1 | 2156 | |||
| BC037887.2 | - | 4 | 3302 | 5274706 | ||
| BC047107.1 | - | 4 | 3135 | 5275217 | ||

KBTBD2
Genomic Map