siRNA Details
| Gene Symbol: SPOUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016390.3 | NP_057474.2 | 11 | 1060 | ||
| XM_017014804.1 | XP_016870293.1 | 12 | 1327 | |||
| GenBank mRNA | AF161459.1 | - | 11 | 1030 | ||
| AL110193.1 | - | 11 | 1060 | |||
| BC010579.1 | - | 11 | 1022 | 4213299 | ||
| BC021273.1 | - | 11 | 1033 | 4843384 | ||
| BC039590.1 | - | 11 | 1045 | 5757547 | ||
| BC046133.1 | - | 11 | 1025 | 5725479 | ||
| BC063644.1 | - | 11 | 1028 | 4539701 | ||

SPOUT1
Genomic Map