siRNA Details
| Gene Symbol: UFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016406.3 | NP_057490.2 | 2 | 415 | ||
| XM_005245254.1 | XP_005245311.1 | 2 | 367 | |||
| XM_017001450.1 | XP_016856939.1 | 3 | 260 | |||
| XM_017001451.1 | XP_016856940.1 | 3 | 524 | |||
| GenBank mRNA | AB154405.1 | - | 2 | 161 | ||
| AF151884.1 | - | 2 | 382 | |||
| AF161504.1 | - | 2 | 401 | |||
| AK292776.1 | - | 2 | 367 | |||
| AK311562.1 | - | 2 | 131 | |||
| BC005187.1 | - | 2 | 407 | 3961569 | ||
| CF128121.1 | - | 2 | 414 | |||

UFC1
Genomic Map