siRNA Details
| Gene Symbol: UFM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286703.1 | NP_001273632.1 | 6 | 1698 | ||
| NM_001286704.1 | NP_001273633.1 | 6 | 1876 | |||
| NM_001286705.1 | NP_001273634.1 | 7 | 1740 | |||
| NM_001286706.1 | NP_001273635.1 | 6 | 2374 | |||
| NM_016617.3 | NP_057701.1 | 6 | 1704 | |||
| NR_104584.1 | 7 | 1765 | ||||
| NR_104585.1 | 5 | 1664 | ||||
| GenBank mRNA | AF208844.1 | - | 6 | 1594 | ||
| AF369953.1 | - | 1 | 646 | |||
| AK095490.1 | - | 6 | 1624 | |||
| CB070175.1 | - | 1 | 150 | |||
| CN362375.1 | - | 1 | 717 | |||

UFM1
Genomic Map