siRNA Details
| Gene Symbol: AGGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018046.4 | NP_060516.2 | 5 | 1168 | ||
| GenBank mRNA | AK289394.1 | - | 1 | 20 | ||
| AW500409.1 | - | 1 | 33 | |||
| AY500994.1 | - | 5 | 1087 | |||
| BC032844.1 | - | 5 | 1154 | 5266978 | ||
| BC073864.1 | - | 5 | 695 | |||
| BE077189.1 | - | 3 | 443 | |||

AGGF1
Genomic Map