siRNA Details
| Gene Symbol: AGGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018046.4 | NP_060516.2 | 6 | 1501 | ||
| GenBank mRNA | AK289394.1 | - | 2 | 353 | ||
| AW500409.1 | - | 2 | 366 | |||
| AY500994.1 | - | 6 | 1420 | |||
| BC015478.1 | - | 1 | 252 | |||
| BC032844.1 | - | 6 | 1487 | 5266978 | ||
| BX648120.1 | - | 2 | 310 | |||

AGGF1
Genomic Map