siRNA Details
| Gene Symbol: AGGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018046.4 | NP_060516.2 | 9 | 1779 | ||
| GenBank mRNA | AK001145.1 | - | 4 | 291 | ||
| AK289394.1 | - | 5 | 631 | |||
| AY500994.1 | - | 9 | 1698 | |||
| BC015478.1 | - | 4 | 530 | |||
| BC029382.1 | - | 2 | 88 | |||
| BC032844.1 | - | 9 | 1765 | 5266978 | ||
| BX648120.1 | - | 5 | 588 | |||

AGGF1
Genomic Map