siRNA Details
| Gene Symbol: FANCI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113378.1 | NP_001106849.1 | 14 | 1438 | ||
| NM_018193.2 | NP_060663.2 | 14 | 1438 | |||
| XM_011521756.2 | XP_011520058.1 | 14 | 1462 | |||
| XM_011521757.2 | XP_011520059.1 | 14 | 1458 | |||
| XM_011521764.2 | XP_011520066.1 | 14 | 1462 | |||
| XM_011521767.2 | XP_011520069.1 | 14 | 1432 | |||
| GenBank mRNA | AB058697.1 | - | 1 | 62 | ||
| AK001581.1 | - | 8 | 827 | |||
| AK027564.1 | - | 3 | 325 | |||
| AK055176.1 | - | 14 | 1397 | |||
| BC140769.1 | - | 14 | 1395 | 9021637 | ||
| BC144483.1 | - | 14 | 1395 | 9053010 | ||
| EF469766.1 | - | 13 | 1348 | |||
| EF567077.1 | - | 13 | 1348 | |||

FANCI
Genomic Map