siRNA Details
| Gene Symbol: CCAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282959.1 | NP_001269888.1 | 14, 15 | 2078 | ||
| NM_001282960.1 | NP_001269889.1 | 14, 15 | 2072 | |||
| NM_018237.3 | NP_060707.2 | 15, 16 | 2117 | |||
| NR_104262.1 | 14, 15 | 1912 | ||||
| GenBank mRNA | AF465616.1 | - | 15, 16 | 2047 | ||
| AK000741.1 | - | 10, 11 | 1431 | |||
| AK001701.1 | - | 15, 16 | 2025 | |||
| AK023438.1 | - | 15, 16 | 2047 | |||
| AK122871.1 | - | 13, 14 | 1892 | |||
| AK292649.1 | - | 15, 16 | 2022 | |||
| AK298004.1 | - | 14, 15 | 1997 | |||
| AK299307.1 | - | 14, 15 | 1977 | |||
| AK307550.1 | - | 10, 11 | 1437 | |||
| AK307671.1 | - | 13, 14 | 1941 | |||
| AK307704.1 | - | 15, 16 | 2022 | |||
| AY249140.1 | - | 15, 16 | 2025 | |||
| BC015475.1 | - | 5, 6 | 656 | |||
| BC026036.2 | - | 14, 15 | 1979 | |||
| BC089420.1 | - | 14, 15 | 1939 | |||
| BC108682.1 | - | 15, 16 | 1995 | |||
| BC130626.1 | - | 15, 16 | 2006 | 40146657 | ||
| BC132725.1 | - | 15, 16 | 2006 | 40146747 | ||
| BC144118.1 | - | 14, 15 | 1801 | |||
| BX647943.1 | - | 13, 14 | 1852 | |||

CCAR1
Genomic Map