siRNA Details
| Gene Symbol: TMEM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018279.3 | NP_060749.2 | 6 | 1844 | ||
| GenBank mRNA | AK223148.1 | - | 6 | 1442 | ||
| BC008596.1 | - | 6 | 1844 | 4154303 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM19| Gene Symbol: TMEM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018279.3 | NP_060749.2 | 6 | 1844 | ||
| GenBank mRNA | AK223148.1 | - | 6 | 1442 | ||
| BC008596.1 | - | 6 | 1844 | 4154303 | ||