siRNA Details
| Gene Symbol: GPALPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316951.1 | NP_001303880.1 | 8 | 1132 | ||
| NM_001316952.1 | NP_001303881.1 | 8 | 1253 | |||
| NM_018559.3 | NP_061029.2 | 8 | 1132 | |||
| XM_005266444.3 | XP_005266501.1 | 8 | 1124 | |||
| XM_011535144.2 | XP_011533446.1 | 8 | 1191 | |||
| GenBank mRNA | AB051491.1 | - | 8 | 1042 | ||
| AF145122.1 | - | 8 | 1067 | |||
| AF245045.1 | - | 7 | 918 | |||
| AK292493.1 | - | 8 | 1105 | |||
| BC032333.1 | - | 8 | 1179 | 5169953 | ||
| BC039586.1 | - | 8 | 1110 | 5722236 | ||

GPALPP1
Genomic Map