siRNA Details
| Gene Symbol: MOSPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306188.1 | NP_001293117.1 | 5 | 945 | ||
| NM_019556.2 | NP_062456.1 | 6 | 1107 | |||
| XM_011531366.2 | XP_011529668.1 | 6 | 1061 | |||
| XM_017029653.1 | XP_016885142.1 | 7 | 1300 | |||
| XM_017029654.1 | XP_016885143.1 | 5 | 899 | |||
| GenBank mRNA | AK299172.1 | - | 4 | 735 | ||
| AK307196.1 | - | 5 | 934 | |||
| AL137163.1 | - | 6 | 1030 | |||
| BC005700.1 | - | 6 | 1010 | 3450155 | ||

MOSPD1
Genomic Map