siRNA Details
| Gene Symbol: IFT80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190241.1 | NP_001177170.1 | 4 | 889 | ||
| NM_001190242.1 | NP_001177171.1 | 2 | 523 | |||
| NM_020800.2 | NP_065851.1 | 3 | 669 | |||
| GenBank mRNA | AB037795.1 | - | 2 | 413 | ||
| AK025109.1 | - | 1 | 579 | |||
| AK055904.1 | - | 4 | 1565 | |||
| AK125342.1 | - | 3 | 669 | |||
| AK303410.1 | - | 3 | 667 | |||
| BC101494.1 | - | 3 | 311 | 8069000 | ||
| BC113669.1 | - | 3 | 311 | 8322721 | ||

IFT80
Genomic Map