siRNA Details
| Gene Symbol: IFT80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190241.1 | NP_001177170.1 | 8, 9 | 1300 | ||
| NM_001190242.1 | NP_001177171.1 | 6, 7 | 934 | |||
| NM_020800.2 | NP_065851.1 | 7, 8 | 1080 | |||
| GenBank mRNA | AB037795.1 | - | 6, 7 | 824 | ||
| AK055904.1 | - | 7, 8 | 1866 | |||
| AK125342.1 | - | 7, 8 | 1080 | |||
| AK303410.1 | - | 7, 8 | 1078 | |||
| BC101494.1 | - | 7, 8 | 722 | 8069000 | ||
| BC113669.1 | - | 7, 8 | 722 | 8322721 | ||

IFT80
Genomic Map