siRNA Details
| Gene Symbol: EPG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020964.2 | NP_066015.2 | 26 | 4592 | ||
| XM_011526122.1 | XP_011524424.1 | 26 | 4592 | |||
| XM_017025889.1 | XP_016881378.1 | 26 | 4574 | |||
| XM_017025890.1 | XP_016881379.1 | 26 | 4574 | |||
| XM_017025891.1 | XP_016881380.1 | 25 | 4451 | |||
| XM_017025892.1 | XP_016881381.1 | 25 | 4617 | |||
| XM_017025893.1 | XP_016881382.1 | 18 | 2464 | |||
| GenBank mRNA | AK023817.1 | - | 10 | 1271 | ||
| BC130614.1 | - | 9 | 1125 | |||
| BC144104.1 | - | 9 | 1125 | |||
| BC144105.1 | - | 9 | 1125 | |||

EPG5
Genomic Map