siRNA Details
| Gene Symbol: BRINP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021165.3 | NP_066988.1 | 4 | 952 | ||
| XM_005245379.1 | XP_005245436.1 | 5 | 1532 | |||
| XM_011509826.1 | XP_011508128.1 | 2 | 779 | |||
| GenBank mRNA | AB051534.1 | - | 4 | 945 | ||
| AB161694.1 | - | 4 | 945 | |||
| AK123336.1 | - | 1 | 387 | |||
| AK296524.1 | - | 4 | 921 | |||
| AL035289.1 | - | 3 | 526 | |||
| BC028036.1 | - | 4 | 844 | 5242899 | ||

BRINP2
Genomic Map